Conexiones de rosca para tubos roscados
Proveedor: LENZ, E. (Lenz Laborglas GmbH & CO.KG)
01321514 01321518 01321525 01321532
LENZ01321514EA 35.6 EUR
LENZ01321514 LENZ01321518 LENZ01321525 LENZ01321532
Conexiones de rosca para tubos roscados
Racores y conectores
Connectors, made of PBT, red with integrated silicone (VMQ)-PTFE sealing.
  • For flexible connection of two DIN glass threads
  • Heat resistant from –45 to +180 °C
  • Extremely leak tight