Nonanoato de metilo, Supelco®
Proveedor: Merck
Sinónimos:
Methyl nonanoate, Nonanoic acid methyl ester, Pelargonic acid methyl ester
M-Clarity™ quality level = MQ100
76368-1ML
76368-5ML
76368-1MLEA
53.2
EUR
76368-1ML
76368-5ML
Nonanoato de metilo, Supelco®
Nonanoato de metilo
Formula:
C₁₀H₂₀O₂ MW: 172,27 g/mol |
Número MDL:
MFCD00009569 Núm. CAS: 1731-84-6 EINECS: 217-052-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...