Magnetic stirring bar retrievers, PTFE coated
Proveedor: KARTELL
710 712
442-4132EA 12.5 EUR
442-4132 442-4131
Magnetic stirring bar retrievers, PTFE coated
Imánes de agitación Herramientas de agitación
PTFE coated stirring bar retrievers for removing magnetic stirrer bars from foodstuff.