D-Treonina 98%

Proveedor: Thermo Scientific

Sinónimos: H-D-Thr-OH, D(+)-Treonina

346970050 346970250 346971000
ACRO346970050EA 48.5 EUR
ACRO346970050 ACRO346970250 ACRO346971000
D-Treonina 98%
D-Treonina
Formula: C₄H₉NO₃
MW: 119,12 g/mol
Pto. de fusión: 274 °C (desc.)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00064269
Núm. CAS: 632-20-2
EINECS: 211-171-8

Order Now

Resultados test especificaciones

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.3 %
Sulfated ash ≤0.1 %
UV transmittance ≥98 % (c=5, H2O, 430 nm) 1 cm cell
Specific optical rotation +27° to +29° (20°C, 589 nm) (c=6, water)
Titration with HClO4 ≥97.5 %
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤300 ppm
Ammonium (NH4) ≤200 ppm
Arsenic (As) ≤1 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Other amino acids (TLC) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR