Triacetato de terbio hidrato ≥99.9% (REO, rare earth oxide basis), REacton®
Proveedor: Thermo Fisher Scientific
Sinónimos:
Terbium(III) acetate hydrate
14600.09
14600.18
14600.09EA
162
EUR
14600.09
14600.18
Triacetato de terbio hidrato ≥99.9% (REO, rare earth oxide basis), REacton®
Triacetato de terbio hidrato
Purity: 99.9%
CAS No.: 100587-92-6
Molecular Formula: C6H9O6Tb.xH2O
Formula Weight: 336.08(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
MDL No.: MFCD00149233
Formula:
C₆H₉O₆Tb·H₂O MW: 354.07 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00149233 Núm. CAS: 100587-92-6 EINECS: 247-065-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...