5-Fenil-o-anísidina ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: 2-Amino-4-phenylanisole, 2-Methoxy-5-phenylaniline, 5-Phenyl-o-anisidine, 3-Amino-4-methoxybiphenyl

A0958-25G A0958-5G
TCIAA0958-25GEA 262 EUR
TCIAA0958-25G TCIAA0958-5G
5-Fenil-o-anísidina ≥98.0% (by titrimetric analysis)
5-Phenyl-o-anisidine
Formula: C₁₃H₁₃NO
MW: 199,25 g/mol
Número MDL: MFCD00007790
Núm. CAS: 39811-17-1
EINECS: 254-639-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7720f1e9770633c6cc3e7c2d26965b01ea1ca015&vid=7720f1e9770633c6cc3e7c2d26965b01ea1ca015&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light red to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 83.0 to 86.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR