Transfer pipettes, Corning® Gosselin™
Proveedor: Corning
PAS1-00 PAS3-00
GOSSPAS1-00EA 26 EUR
GOSSPAS1-00 GOSSPAS3-00
Transfer pipettes, Corning® Gosselin™
Pipetas Pipetas de transferencia (Pasteur)
Disposable, sterile pipettes made of PE.
  • Graduated
  • Non pyrogenic, non cytotoxic, non haemolytic
  • Product traceability on each bag
  • Sterility Assurance Level based on ISO 11137
Typical application areas include: Dairy, environment, industries, microbiology.