Tubing connectors, T-shaped, LaboPlast®
Proveedor: BURKLE
8705-0014 8705-0006 8705-0012 8705-0004 8711-0004 8705-0008 8711-0008 8711-0010 8711-0006 8711-0012 8711-0014 8705-0010
229-0127EA 16.9 EUR
229-0127 229-0124 229-0126 229-0123 229-0132 229-0125 229-0147 229-0148 229-0146 229-0149 229-0199 BURK8705-0010
Tubing connectors, T-shaped, LaboPlast®
Racores y conectores
PP or PVDF, translucent.
  • PP, temperature resistant up to +135 °C max.
  • PVDF, temperature resistant up to +160 °C max., highly resistant to chemicals
  • Sterilisable
Cylindrical T hose connector to connect hoses. PVDF connectors with food industry approval, autoclavable and UV-resistant.