3-Metilfenetilamina 98%
Proveedor: Thermo Fisher Scientific
|
Peligro
|
351110010
351110050
ACRO351110010EA
35.1
EUR
ACRO351110010
ACRO351110050
3-Metilfenetilamina 98%
3-Metilfenetilamina
Formula:
C₉H₁₃N Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD01310828 Núm. CAS: 55755-17-4 UN: 2735 ADR: 8,III |
Resultados test especificaciones
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...