Trifluorometanosulfonato de potasio ≥98%
Proveedor: Thermo Fisher Scientific
|
Atención
|
20361.09
20361.18
20361.09EA
70.2
EUR
20361.09
20361.18
Trifluorometanosulfonato de potasio ≥98%
Trifluorometanosulfonato de potasio
Purity: 98%
CAS No.: 2926-27-4
Molecular Formula: CF3SO3K
Formula Weight: 188.17
Physical Form: Crystalline
Sensitivity: Hygroscopic
MDL No.: MFCD00042370
Formula:
CF₃KO₃S MW: 188.17 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00042370 Núm. CAS: 2926-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...