2-Picolil cloruro clorhidrato ≥97.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

C0822-250G C0822-25G
TCIAC0822-250GEA 400 EUR
TCIAC0822-250G TCIAC0822-25G
2-Picolil cloruro clorhidrato ≥97.0% (by titrimetric analysis)
2-Picolil cloruro clorhidrato
Formula: C₆H₇Cl₂N
Número MDL: MFCD00012811
Núm. CAS: 6959-47-3
EINECS: 230-149-9
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36af69a5bad6a7caee38bb92edb62625549c5269&vid=36af69a5bad6a7caee38bb92edb62625549c5269&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 124.0 to 127.0 deg-C
Solubility in Water almost transparency

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR