Potasio hidrogenocarbonato 99.5-101.0% (by acidimetry), Puriss., meets analytical specification of BP, Ph. Eur., USP, E501, Fluka™

Proveedor: Honeywell Chemicals

Sinónimos: Potasio bicarbonato

12602-25KG 12602-1KG
12602-25KGEA 389 EUR
12602-25KG 12602-1KG
Potasio hidrogenocarbonato 99.5-101.0% (by acidimetry), Puriss., meets analytical specification of BP, Ph. Eur., USP, E501, Fluka™
Potasio hidrogenocarbonato
Formula: KHCO₃
Pto. de fusión: 100-120 °C (desc.)
Densidad: 2,17 g/cm³ (20 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011402
Núm. CAS: 298-14-6
EINECS: 206-059-0
Índice Merck: 13,07691

Order Now

Resultados test especificaciones

Assay (acidimetric) 99.5 - 101.0 %
Assay (calculated to the dried substance) 99.5 - 101 %
Loss on drying (on silicagel, 4h) Max. 0.1 %
pH (5 %, 20°C) 8.0 - 8.6
Ammonium (NH₄) Max. 20 ppm
Arsenic (As) Max. 1 ppm
Calcium (Ca) Max. 50 ppm
Iron (Fe) Max. 10 ppm
Mercury (Hg) Max. 1 ppm
Sodium (Na) Max. 0.03 %
Lead (Pb) Max. 2 ppm
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 50 ppm
Carbonate (K₂CO₃) Max. 2.5 %
Sulfate (SO₄) Max. 100 ppm
Residual solvents complying
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR