Potasio yoduro ≥99.0% ACS
Proveedor: Thermo Fisher Scientific
Sinónimos:
Dipotassium diiodide, Kali iodide
11601.22
11601.36
11601.A3
11601.22EA
59.9
EUR
11601.22
11601.36
11601.A3
Potasio yoduro ≥99.0% ACS
Potasio yoduro
Purity: 99.0% min
CAS No.: 7681-11-0
Molecular Formula: KI
Formula Weight: 166.01
Physical Form: Granular
Density: 3.12
Refractive Index: 1.677
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 681°
Boiling Point: 1330°
Solubility: Soluble in water, alcohol, acetone, and glycerol
Merck Reference: 14,7643
MDL No.: MFCD00011405
In photographic emulsions, in analytical chemistry, as a feed additive, in spectroscopy, in infrared transmission, and in scintillation
Formula:
Kl MW: 166 g/mol Pto. Ebullición: 1330 °C Pto. de fusión: 681 °C Densidad: 3.12 Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00011405 Núm. CAS: 7681-11-0 EINECS: 213-659-4 Índice Merck: 14,7643 |