Silicagel, Pore size: 60 Å 0.062-0.105 mm (150-230 mesh)
Proveedor: Thermo Fisher Scientific
|
Atención
|
42727.30
42727.A1
42727.A7
42727.30EA
89.1
EUR
42727.30
42727.A1
42727.A7
Silicagel, Pore size: 60 Å 0.062-0.105 mm (150-230 mesh)
Silicagel
CAS No.: 7631-86-9
Physical Form: -150+230 Mesh, S.A. 500-600m2/g
Odour: Odourless
MDL No.: MFCD00011232
Column chromatography without applied pressure (gravity pulled)
Formula:
SiO₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00011232 Núm. CAS: 63231-67-4 EINECS: 231-545-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...