Diseleniuro de volframio ≥99.8% (metals basis)
Proveedor: Thermo Fisher Scientific
|
Peligro
|
13084.09
13084.18
13084.09EA
134
EUR
13084.09
13084.18
Diseleniuro de volframio ≥99.8% (metals basis)
Diseleniuro de volframio
Purity: 99.8%
CAS No.: 12067-46-8
Molecular Formula: WSe2
Formula Weight: 341.77
Physical Form: 10-20 Micron powder
Density: 9.32
Appearance: Black
Odour: Odourless
UN No.: 3077
MDL No.: MFCD00049703
Formula:
Se₂W MW: 341.76 g/mol Densidad: 9.32 Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00049703 Núm. CAS: 12067-46-8 EINECS: 235-078-7 UN: 3077 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...