Industrial gloves, Polytril™ Air Comfort
Proveedor: HONEYWELL SAFETY
2232273/7 2232273/8 2232273/9 2232273/10 2232273/11
112-1539EA 70.1 EUR
112-1539 112-1540 112-1541 112-1537 112-1538
Industrial gloves, Polytril™ Air Comfort
Guantes
Polyamide/cotton/elasthane knitted glove with black foam nitrile coating on palm and fingers, an open back and an elasticated knitted wrist.
  • Ergonomic seamless knit construction conforms better to the natural shape of the hand for greater strength and durability
  • Lycra® yarn gives a better tactile sensitivity for the assembling of small parts
  • Addition of cotton increases the absorption of perspiration and comfort
  • Nitrile foam coating and open back provide oil and abrasion resistance, improved grip and superior ventilation
  • Skin is not in direct contact with the coating, which helps keeping hands cool and dry
Suitable for light handling and assembling in an oily environment, as well as handling in a wet and greasy environment. Also ideal for assembly work, plumbing, joinery, construction, automotive industry and the handling of white and brown goods.
Certificaciones: EN 388:2016 (3121X); Cat. II