Tubing connectors, T-shaped, BOLA
Proveedor: Bohlender
D577-06 D577-08 D577-04 D577-02
BOHLD577-06EA 34.8 EUR
BOHLD577-06 BOHLD577-08 BOHLD577-04 BOHLD577-02
Tubing connectors, T-shaped, BOLA
Racores y conectores
Tubing connectors, T-shaped, made of PTFE, for flexible tubings.
  • Universal chemical resistance
  • Temperature resistance: from −200 to +250 °C
  • Sutable for vacuum applications
  • FDA conform