2-Amino-1,3,4-tiadiazol ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: [1,3,4]Thiadiazol-2-ylamine, 1,3,4-Thiadiazol-2-amine

A1060-25G A1060-5G
TCIAA1060-25GEA 102 EUR
TCIAA1060-25G TCIAA1060-5G
2-Amino-1,3,4-tiadiazol ≥98.0% (by HPLC, titration analysis)
2-Amino-1,3,4-thiadiazole
Formula: C₂H₃N₃S
MW: 101,13 g/mol
Número MDL: MFCD00003107
Núm. CAS: 4005-51-0
EINECS: 223-657-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=759bb9ee4eb81edca8870e997b6762ff7ec54c6f&vid=759bb9ee4eb81edca8870e997b6762ff7ec54c6f&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR