Single use syringes for insulin injection, Omnican®
Proveedor: B. Braun
9151133S 9151117S 9151125S
720-2556EA 72.5 EUR
720-2556 BRAU9151117S BRAU9151125S
Single use syringes for insulin injection, Omnican®
Jeringas Jeringas médicas
Latex free single use syringe with integrated cannula for subcutaneous injection of U-100 insulin (50 or 100 IU/ml).
  • Plunger stopper prevents the plunger from coming out of the syringe cylinder, ensuring medication is not wasted
  • Large, easy to read scale enables reliable dosage, air bubbles can be easily seen through the clear body of the syringe
  • Protective caps at the ends of the syringe ensures sterility
  • All syringes are also available with a short 8 mm cannulae to avoid unintentional injection into muscle
  • Precise, triple facet bevel and silicone coated cannulae means the puncture is barely noticeable
Envase: 10×10 units in a polybag.