Reducing screw thread adaptor couplings, BOLA
Proveedor: Bohlender
H904-06 H904-07 H904-02 H904-03 H904-04 H904-08 H904-05
BOHLH904-06EA 60.5 EUR
BOHLH904-06 BOHLH904-07 BOHLH904-02 BOHLH904-03 BOHLH904-04 BOHLH904-08 BOHLH904-05
Reducing screw thread adaptor couplings, BOLA
Racores y conectores
Connector, reducing screw thread coupling, made of PTFE. Two screw caps made of PPS with GL thread and a PTFE/FPM gasket. The product is only exposed to PTFE.
  • Applicable for connecting two GL threads of different sizes
  • Temperature resisitance from −200 to +250 ºC
  • Very good chemical resistance
  • FDA conformity