Corning® Vitronectin, human, 0,25 mg, is used to promote cell attachment, spreading, proliferation, and differentiation of many normal and neoplastic cells, and to study cell migration.
Control de calidad mediante SDS-PAGE, análisis y obtención de resultados negativos para bacterias, hongos y micoplasma Análisis del material original para la detección del antígeno de la hepatitis B y el anticuerpo anti-VIH-1
Molecular Weight 75 kD and 65 kD components. Stable for at least three months at 2° to 8°C (lyophilized) or one week at 2° to 8°C (solubilized). Recommended concentration: 50 ng/cm2 of growth surface depending on cell type. formulation is Lyophilized dialyzed against 10 mM phosphate buffer pH 7.7.