7-Bromo-1,2,3,4-tetrahidronaftalen-1-ona
Proveedor: Cayman Chemical
Sinónimos:
7-Bromo-3,4-dihydronaphthalen-1(2H)-one
10007135-100
10007135-250
10007135-50
10007135-500
CAYM10007135-100EA
48.7
EUR
CAYM10007135-100
CAYM10007135-250
CAYM10007135-50
CAYM10007135-500
7-Bromo-1,2,3,4-tetrahidronaftalen-1-ona
7-Bromo-1,2,3,4-tetrahidronaftalen-1-ona
Formula:
C₁₀H₉BrO MW: 225,08 g/mol |
Número MDL:
MFCD02179287 Núm. CAS: 32281-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...