Heptafluorotantalato de dipotasio ≥99.7%
Proveedor: Thermo Scientific
|
Peligro
|
Sinónimos:
Potassium heptafluorotantalate(V)
40316.18
40316.30
40316.18EA
364
EUR
40316.18
40316.30
Heptafluorotantalato de dipotasio ≥99.7%
Heptafluorotantalato de dipotasio
Purity: 99.7%
CAS No.: 16924-00-8
Molecular Formula: K2TaF7
Formula Weight: 392.14
Physical Form: Powder
Density: 4.560
Solubility: Decomposes in water. Slightly soluble in HF
UN No.: 3288
MDL No.: MFCD00042369
Formula:
F₇K₂Ta MW: 392,13 g/mol Densidad: 4,560 Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00042369 Núm. CAS: 16924-00-8 EINECS: 240-986-1 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...