Glicinato de terc-butilo clorhidrato ≥98.0% (by total nitrogen and titration analysis)

Proveedor: TCI

Sinónimos: H-Gly-OtBu.HCl, Glycine tert-butyl ester hydrochloride, H-Gly-OtBu·HCl

G0254-1G G0254-25G G0254-5G
TCIAG0254-1GEA 17 EUR
TCIAG0254-1G TCIAG0254-25G TCIAG0254-5G
Glicinato de terc-butilo clorhidrato ≥98.0% (by total nitrogen and titration analysis)
Glicinato de terc-butilo clorhidrato
Formula: C₆H₁₄ClNO₂
MW: 167,64 g/mol
Número MDL: MFCD00058255
Núm. CAS: 27532-96-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e4627e012a974078c3256c59e330c63e34c5929&vid=7e4627e012a974078c3256c59e330c63e34c5929&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR