N-Fenil-N-nitrosohidroxilamina sal amonica

Proveedor: Thermo Fisher Scientific
Peligro

Sinónimos: Ammonium 2-oxo-1-phenylhydrazinolate

423601000 423605000 423600250
ACRO423601000EA 73.5 EUR
ACRO423601000 ACRO423605000 ACRO423600250
N-Fenil-N-nitrosohidroxilamina sal amonica
N-Fenil-N-nitrosohidroxilamina sal amonica
Formula: C₆H₉N₃O₂
Pto. de fusión: 150…155 °C (desc.)
Temperatura de almacenaje: Nevera
Número MDL: MFCD00078422
Núm. CAS: 135-20-6
EINECS: 205-183-2
UN: 2811
ADR: 6.1,III

Order Now

Resultados test especificaciones

Appearance White to yellow-brown Crystalline powder or flakes
Infrared spectrum Conforms
Additional info package usually contains small pouches of ammonium carbonate as preservative
Solubility (5 % in water) Passes test
Residue after ignition ≤0.05 %
Suitability (for precip. of Fe, etc.) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR