Ectoina
Proveedor: Merck
Sinónimos:
(S)-2-Methyl-3,4,5,6-tetrahydropyrimidine-4-carboxylic acid
1.30200.1001
1.30200.0100
1.30200.5001
1.30200.1001EA
0
EUR
1.30200.1001
1.30200.0100
1.30200.5001
Ectoina
Ectoina
Formula:
C₆H₁₀N₂O₂ MW: 142,1558 g/mol |
Número MDL:
MFCD03419286 Núm. CAS: 96702-03-3 EINECS: 431-910-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...