Welding jacket with waistband, Flam
Proveedor: FRISTADS KANSAS
1003345402XL 100334540L 100334540M 100334540S 100334540XL 100334540XS
113-3909EA 155.9 EUR
113-3909 113-3910 113-3911 113-3912 113-3908 113-3913
Welding jacket with waistband, Flam
Ropa de trabajo
The fabric of this jacket is made up of 75% cotton and 25% polyester and has been treated with the flame-retardant PROBAN®. Thanks to this treatment the jacket protects against flames, welding sparks and open arcs.
  • Concealed zip and buttons up to the top of the collar
  • Two breast and side pockets with flap and concealed press studs, inner pocket with mobile phone pouch and loop for earphones
  • Adjustable cuffs with rip-tape fastening, adjustable bottom edge with buttons
Weight: 365 g/m²
EN 61482-1-2 (class 1), EN ISO 11611 (A1 class 1), EN ISO 11612 (More Product Information
Order Now
ESPECIFICACIONES
Modelo FLAM-4031
Environmentally Preferable
Aplicaciones de vestimenta Welding
Risk area Heat,Flame-retardant
Género Para hombres
Sleeve or leg length Long
Tamaño de cierre Con cremallera
Tela 75% cotton, 25% polyester
Peso 365 g/m²