Cation exchange chromatography columns, IEC series
Proveedor: Resonac
F6110002 F6118250
554-2851EA 1076 EUR
554-2851 554-2856
Cation exchange chromatography columns, IEC series
Columnas de cromatografía
SP-825, CM-825: Suitable for the analysis of relatively high molecular weight compounds, such as proteins, peptides, DNA, and RNA. Usable across wide pH range between 2 and 12.
SP-420N: Non porous base material. For rapid analysis.