D-Gluconato de cinc hidrato ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

G0277-25G G0277-500G
TCIAG0277-25GEA 16 EUR
TCIAG0277-25G TCIAG0277-500G
D-Gluconato de cinc hidrato ≥98.0% (by titrimetric analysis)
D-Gluconato de cinc hidrato
Formula: C₁₂H₂₂O₁₄Zn·xH₂O
Número MDL: MFCD04037230
Núm. CAS: 4468-02-4
EINECS: 224-736-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab7987aa349cc83d61013f94b211bbd30a6bb7ed&vid=af694336e2eef7ffc895de6b36e4d923e08f1251&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +8.0 to +11.0 deg (C=1, H2O)
Solubility in Water almost transparency
Water max. 11.0 %

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR