Thionin(acetate)

Proveedor: Thermo Fisher Scientific
Atención

Sinónimos: 3,7-Diaminophenothiazin-5-ium acetate

A18912.06 A18912.14
A18912.06EA 63.6 EUR
A18912.06 A18912.14
Thionin(acetate)
Thionin(acetate)

CAS No.: 78338-22-4 Molecular Formula: C14H13N3O2S Formula Weight: 287.34 MDL No.: MFCD00081194


Used as a biogical stain

Formula: C₁₄H₁₃N₃O₂S
MW: 287.34 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00081194
Núm. CAS: 78338-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR