4-[Bis(p-hidroxifenil)metileno]ciclohexa-2,5-dien-1-ona

Proveedor: Thermo Fisher Scientific
Atención

Sinónimos: 4-(Bis(4-hydroxyphenyl)methylene)cyclohexa-2,5-dien-1-one

229830250 229831000 229835000
ACRO229830250EA 85.7 EUR
ACRO229830250 ACRO229831000 ACRO229835000
4-[Bis(p-hidroxifenil)metileno]ciclohexa-2,5-dien-1-ona
4-[Bis(p-hidroxifenil)metileno]ciclohexa-2,5-dien-1-ona
Formula: C₁₉H₁₄O₃
MW: 290.32 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00001624
Núm. CAS: 603-45-2
EINECS: 210-041-8

Order Now

Resultados test especificaciones

Appearance Red-brown to brown Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤10 % (110°C, 1 h)
UV Lambda max 480 to 486 nm (in ethanol + 2ml 1N HCl)
Visual transition interval from pH 6.2 (yellow) to pH 8.2 (red-pink)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR