1-Cloroftalazina ≥97%
Proveedor: Thermo Scientific
|
Atención
|
H27143.09
H27143.03
H27143.09EA
408
EUR
H27143.09
H27143.03
1-Cloroftalazina ≥97%
1-Cloroftalazina
Purity: 97%
CAS No.: 5784-45-2
Molecular Formula: C8H5N2Cl
Formula Weight: 164.59
Storage Temperature: 2°C to 8°C
Melting Point: 109-112°
MDL No.: MFCD00024141
Formula:
C₈H₅ClN₂ MW: 164,59 g/mol Pto. de fusión: 109…112 °C |
Número MDL:
MFCD00024141 Núm. CAS: 5784-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...