Glove liners, Perfect Poly® PU First Liner White
Proveedor: HONEYWELL SAFETY
2132200/10 2132200/7 2132200/8 2132200/9
DALL2132200/10EA 120 EUR
DALL2132200/10 DALL2132200/7 DALL2132200/8 DALL2132200/9
Glove liners, Perfect Poly® PU First Liner White
Guantes
50% polyamide, 50% polyester knitted glove for fine handling or for use as an underglove.
  • Knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
  • Elasticated wrist
  • Non pilling, the polyamide is suitable for precision work
  • White colour enhances the glove's hygiene and cleanliness
Certificaciones: Cat. I