Bottles, narrow or wide neck, soda-lime glass, clear or amber, with standard ground joint
Proveedor: GLASWARENFABRIK KARL HECHT
41301018 41301006 41303024 41311012 41311024 41311030
HECH41301018EA 10.7 EUR
HECH41301018 HECH41301006 HECH41303024 HECH41311012 HECH41311024 HECH41311030
Bottles, narrow or wide neck, soda-lime glass, clear or amber, with standard ground joint
Frascos Botellas de vidrio
Bottles, made of soda-lime glass, with ground-in flat glass stopper.
  • Good chemical and physical properties
  • Not thermal shock-resistant
  • Solid glass stoppers