3-Metoxitiobenzamida

Proveedor: Cayman Chemical

10553-500 10553-1 10553-5
CAYM10553-500EA 29.7 EUR
CAYM10553-500 CAYM10553-1 CAYM10553-5
3-Metoxitiobenzamida
3-Metoxitiobenzamida
Formula: C₈H₉NOS
Número MDL: MFCD04627361
Núm. CAS: 64559-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR