Ácido 5-metilpirazin-2-carboxílico ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Peligro
|
M1003-25G
M1003-5G
TCIAM1003-25GEA
60
EUR
TCIAM1003-25G
TCIAM1003-5G
Ácido 5-metilpirazin-2-carboxílico ≥98.0% (by HPLC, titration analysis)
Ácido 5-metilpirazin-2-carboxílico
Formula:
C₆H₆N₂O₂ MW: 138.13 g/mol |
Número MDL:
MFCD00068241 Núm. CAS: 5521-55-1 EINECS: 413-260-9 |
|
Resultados test especificaciones
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...