1-(Trifenilfosforaniliden)acetona ≥99%
Proveedor: Thermo Fisher Scientific
Sinónimos:
1-(Triphenylphosphoranylidene)propan-2-one, 1-Triphenylphosphoranylidene-2-propanone
A11410.09
A11410.18
A11410.30
A11410.09EA
57.3
EUR
A11410.09
A11410.18
A11410.30
1-(Trifenilfosforaniliden)acetona ≥99%
1-(Trifenilfosforaniliden)acetona
Purity: 99%
CAS No.: 1439-36-7
Molecular Formula: C21H19OP
Formula Weight: 318.36
Storage Temperature: 2°C to 8°C
Melting Point: 204-209°
Beilstein Reference: 750077
MDL No.: MFCD00008774
Formula:
C₂₁H₁₉OP Pto. de fusión: 204…209 °C |
Número MDL:
MFCD00008774 Núm. CAS: 1439-36-7 EINECS: 215-878-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...