Razor blades, Gem®
Proveedor: PERSONNA MEDICAL
62-0325 62-0335-0000
PERS62-0325EA 2410 EUR
PERS62-0325 PERS62-0335-0000
Razor blades, Gem®
Cuchillas
Razor blades for catheter cutting or cleanroom applications.
  • High precision, sharpness and safety
  • Made from carbon steel or from stainless steel
Carbon steel blades are suitable for ISO Class 7 cleanroom environments.
Stainless steel blades are suitable for ISO Class 5, 6 and 7 cleanroom environments.