Ácido 2-bromo-3-nitrobenzoico ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
B3322-25G
B3322-5G
TCIAB3322-25GEA
505
EUR
TCIAB3322-25G
TCIAB3322-5G
Ácido 2-bromo-3-nitrobenzoico ≥98.0% (by titrimetric analysis)
Ácido 2-bromo-3-nitrobenzoico
Formula:
C₇H₄BrNO₄ MW: 246,02 g/mol |
Número MDL:
MFCD00074899 Núm. CAS: 573-54-6 EINECS: 209-356-3 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 186.0 to 191.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...