2-Cyanobenzaldehído ≥99.0% (por GC)

Proveedor: TCI
Peligro

Sinónimos: 2-Formylbenzonitrile

C0444-25G C0444-5G
TCIAC0444-25GEA 299 EUR
TCIAC0444-25G TCIAC0444-5G
2-Cyanobenzaldehído ≥99.0% (por GC)
2-Cyanobenzaldehído
Formula: C₈H₅NO
MW: 131,13 g/mol
Número MDL: MFCD00017503
Núm. CAS: 7468-67-9
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c32b14b32167efd6acacd551e86468d39ff8247&vid=5c32b14b32167efd6acacd551e86468d39ff8247&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 102.0 to 109.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR