Ácido 2-cloro-m-toluico ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

C2114-25G C2114-5G
TCIAC2114-25GEA 117 EUR
TCIAC2114-25G TCIAC2114-5G
Ácido 2-cloro-m-toluico ≥98.0% (by GC, titration analysis)
Ácido 2-cloro-m-toluico
Formula: C₈H₇ClO₂
MW: 170,6 g/mol
Número MDL: MFCD00079736
Núm. CAS: 15068-35-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8550891252df64fb0685684ff53342d01731318f&vid=8550891252df64fb0685684ff53342d01731318f&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 140.0 to 144.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR