3-Amino-6-cloropiridazina ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
6-Cloropiridazin-3-amina
A1817-25G
A1817-5G
TCIAA1817-25GEA
48
EUR
TCIAA1817-25G
TCIAA1817-5G
3-Amino-6-cloropiridazina ≥98.0% (by GC, titration analysis)
3-Amino-6-cloropiridazina
Formula:
C₄H₄ClN₃ MW: 129,55 g/mol |
Número MDL:
MFCD00051506 Núm. CAS: 5469-69-2 EINECS: 226-796-1 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Acetic acid | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...