4-Amino-3-fluorofenol ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

A2164-1G A2164-5G
TCIAA2164-1GEA 49 EUR
TCIAA2164-1G TCIAA2164-5G
4-Amino-3-fluorofenol ≥98.0% (by GC, titration analysis)
4-Amino-3-fluorophenol
Formula: C₆H₆FNO
MW: 127,12 g/mol
Número MDL: MFCD00077452
Núm. CAS: 399-95-1
EINECS: 402-230-0
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9bec6d37e19f8e9a5ed27cfd254c11581787b54&vid=a9bec6d37e19f8e9a5ed27cfd254c11581787b54&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 135.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR