Tetrabutilamonio ioduro ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Ioduro de tetrabutilamonio
T0057-25G
T0057-500G
T0057-100G
TCIAT0057-25GEA
31
EUR
TCIAT0057-25G
TCIAT0057-500G
TCIAT0057-100G
Tetrabutilamonio ioduro ≥98.0% (by titrimetric analysis)
Tetrabutilamonio ioduro
Formula:
C₁₆H₃₆IN MW: 369.37 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00011636 Núm. CAS: 311-28-4 EINECS: 206-220-5 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystaline |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 144.0 to 149.0 deg-C |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...