4-Bromo-2,3-difluorofenol ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
D3913-1G
D3913-5G
TCIAD3913-1GEA
16
EUR
TCIAD3913-1G
TCIAD3913-5G
4-Bromo-2,3-difluorofenol ≥98.0% (by GC, titration analysis)
4-Bromo-2,3-difluorofenol
Formula:
C₆H₃BrF₂O |
Número MDL:
MFCD08061907 Núm. CAS: 144292-32-0 |
|
Resultados test especificaciones
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 53.0 to 57.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...