3-Cloro-6-metilpiridazina ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
C2274-25G
C2274-5G
TCIAC2274-25GEA
282
EUR
TCIAC2274-25G
TCIAC2274-5G
3-Cloro-6-metilpiridazina ≥98.0% (by GC, titration analysis)
3-Cloro-6-metilpiridazina
Formula:
C₅H₅ClN₂ MW: 128,56 g/mol |
Número MDL:
MFCD00052905 Núm. CAS: 1121-79-5 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 58.0 to 62.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...