trans-2,4-Dichlorocinnamic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: (E)-3-(2,4-Dichlorophenyl)acrylic acid, (2E)-3-(2,4-Dichlorophenyl)acrylic acid

D2588-25G D2588-5G
TCIAD2588-25GEA 133 EUR
TCIAD2588-25G TCIAD2588-5G
trans-2,4-Dichlorocinnamic acid ≥98.0% (by GC, titration analysis)
trans-2,4-Dichlorocinnamic acid
Formula: C₉H₆Cl₂O₂
MW: 217,05 g/mol
Número MDL: MFCD00004373
Núm. CAS: 20595-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6a7672622f986e53022124a11ab8ad60e8ed3a&vid=8b6a7672622f986e53022124a11ab8ad60e8ed3a&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 234.0 to 238.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR