4'-Fluorochalcona ≥98.0% (by HPLC)

Proveedor: TCI
Atención

Sinónimos: 1-(4-Fluorophenyl)-3-phenylprop-2-en-1-one, 4'-Fluorobenzylidenacetophenone

F0425-25G F0425-5G
TCIAF0425-25GEA 113 EUR
TCIAF0425-25G TCIAF0425-5G
4'-Fluorochalcona ≥98.0% (by HPLC)
4'-Fluorochalcona
Formula: C₁₅H₁₁FO
MW: 226,25 g/mol
Número MDL: MFCD00017961
Núm. CAS: 399-10-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3856d5df24fd3c80765ee115874e1b048d635f7&vid=b3856d5df24fd3c80765ee115874e1b048d635f7&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 78.0 to 81.0 deg-C
Solubility in Acetonitrile almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR