4-Amino-2-fluorofenol ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

A2163-1G A2163-5G
TCIAA2163-1GEA 48 EUR
TCIAA2163-1G TCIAA2163-5G
4-Amino-2-fluorofenol ≥98.0% (by GC, titration analysis)
4-Amino-2-fluorofenol
Formula: C₆H₆FNO
MW: 127.12 g/mol
Número MDL: MFCD00671760
Núm. CAS: 399-96-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9fee6f982f5c1827464584de640337e1bf74155&vid=d9fee6f982f5c1827464584de640337e1bf74155&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR