Ácido (3-iodofenil)acético ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-(3-Iodophenyl)acetic acid
I0760-1G
I0760-5G
TCIAI0760-1GEA
20
EUR
TCIAI0760-1G
TCIAI0760-5G
Ácido (3-iodofenil)acético ≥98.0% (by GC, titration analysis)
Ácido (3-iodofenil)acético
Formula:
(C₆H₄I)CH₂COOH MW: 262,05 g/mol |
Número MDL:
MFCD00046548 Núm. CAS: 1878-69-9 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 129.0 to 133.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...